Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009998.3 | NP_001009998.1 | 13 | 1106 | ||
NM_032627.4 | NP_116016.1 | 14 | 1172 | |||
XM_005259790.3 | XP_005259847.1 | 14 | 1213 | |||
XM_006722665.3 | XP_006722728.1 | 14 | 1219 | |||
XM_006722666.2 | XP_006722729.1 | 14 | 1177 | |||
XM_006722668.2 | XP_006722731.1 | 13 | 1111 | |||
XM_017026431.1 | XP_016881920.1 | 14 | 1231 | |||
XM_017026432.1 | XP_016881921.1 | 14 | 1769 | |||
XM_017026433.1 | XP_016881922.1 | 14 | 1177 | |||
XM_017026434.1 | XP_016881923.1 | 13 | 1160 | |||
XM_017026435.1 | XP_016881924.1 | 13 | 1702 | |||
XM_017026436.1 | XP_016881925.1 | 13 | 1111 | |||
XM_017026437.1 | XP_016881926.1 | 13 | 1142 | |||
GenBank mRNA | BC000274.1 | - | 14 | 1079 | 3356283 |