Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114397.1 | NP_001107869.1 | 14 | 1674 | ||
NM_020865.2 | NP_065916.2 | 14 | 1716 | |||
GenBank mRNA | AB040921.1 | - | 12 | 1168 | ||
AF217190.1 | - | 14 | 1708 | |||
AJ577133.1 | - | 14 | 1635 | |||
AJ577134.1 | - | 14 | 1593 | |||
AK303720.1 | - | 1596 | ||||
AK314435.1 | - | 14 | 1716 | |||
BC036035.1 | - | 14 | 1680 | 5273384 |