Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114397.1 | NP_001107869.1 | 25 | 2943 | ||
NM_020865.2 | NP_065916.2 | 25 | 2985 | |||
GenBank mRNA | AB040921.1 | - | 23 | 2437 | ||
AF217190.1 | - | 25 | 2977 | |||
AJ577133.1 | - | 25 | 2904 | |||
AJ577134.1 | - | 25 | 2862 | |||
AK314435.1 | - | 25 | 2985 | |||
BC036035.1 | - | 24 | 2862 | 5273384 |