Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287392.1 | NP_001274321.1 | 3 | 380 | ||
NM_001287393.1 | NP_001274322.1 | 3 | 365 | |||
NM_001287394.1 | NP_001274323.1 | 2 | 196 | |||
NM_203495.3 | NP_987091.1 | 3 | 200 | |||
NM_203497.3 | NP_987093.1 | 3 | 200 | |||
XM_011534948.2 | XP_011533250.1 | 4 | 293 | |||
GenBank mRNA | AF169971.1 | - | 3 | 167 | ||
AK000009.1 | - | 3 | 169 | |||
AY542161.1 | - | 3 | 133 | |||
BC110837.1 | - | 3 | 137 | |||
BC117391.1 | - | 3 | 177 | 40125942 | ||
BC143912.1 | - | 3 | 177 | 9052430 | ||
BC171758.1 | - | 3 | 177 | 9054411 | ||
BM476717.1 | - | 133 | ||||
BX538116.1 | - | 3 | 323 | |||
CD175553.1 | - | 3 | 200 | |||
DB487154.2 | - | 3 | 183 | |||
DB489071.1 | - | 3 | 373 | |||
DB509325.2 | - | 2 | 185 | |||
HY028175.1 | - | 3 | 173 | |||
HY045286.1 | - | 3 | 310 | |||
HY105279.1 | - | 3 | 201 |