Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287392.1 | NP_001274321.1 | 3 | 353 | ||
NM_001287393.1 | NP_001274322.1 | 3 | 338 | |||
NM_001287394.1 | NP_001274323.1 | 2 | 169 | |||
NM_203495.3 | NP_987091.1 | 3 | 173 | |||
NM_203497.3 | NP_987093.1 | 3 | 173 | |||
XM_011534948.2 | XP_011533250.1 | 4 | 266 | |||
GenBank mRNA | AF169971.1 | - | 3 | 140 | ||
AK000009.1 | - | 3 | 142 | |||
AY542161.1 | - | 3 | 106 | |||
BC110837.1 | - | 3 | 110 | |||
BC117391.1 | - | 3 | 150 | 40125942 | ||
BC143912.1 | - | 3 | 150 | 9052430 | ||
BC171758.1 | - | 3 | 150 | 9054411 | ||
BM476717.1 | - | 106 | ||||
BX538116.1 | - | 3 | 296 | |||
CD175553.1 | - | 3 | 173 | |||
DB487154.2 | - | 3 | 156 | |||
DB489071.1 | - | 3 | 346 | |||
DB509325.2 | - | 2 | 158 | |||
HY028175.1 | - | 3 | 146 | |||
HY045286.1 | - | 3 | 283 | |||
HY105279.1 | - | 3 | 174 |