Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139057.3 | NP_620688.2 | 13, 14 | 1988 | ||
XM_005254872.3 | XP_005254929.1 | 13, 14 | 2904 | |||
XM_011521312.2 | XP_011519614.1 | 13, 14 | 2904 | |||
XM_017021973.1 | XP_016877462.1 | 14, 15 | 3037 | |||
XM_017021974.1 | XP_016877463.1 | 14, 15 | 3039 | |||
XM_017021975.1 | XP_016877464.1 | 14, 15 | 3037 | |||
XM_017021976.1 | XP_016877465.1 | 13, 14 | 1749 | |||
XM_017021977.1 | XP_016877466.1 | 14, 15 | 3036 | |||
XM_017021978.1 | XP_016877467.1 | 7, 8 | 4295 | |||
XM_017021979.1 | XP_016877468.1 | 6, 7 | 745 | |||
XM_017021980.1 | XP_016877469.1 | 6, 7 | 955 | |||
XM_017021981.1 | XP_016877470.1 | 14, 15 | 3034 | |||
XM_017021982.1 | XP_016877471.1 | 4, 5 | 565 | |||
XM_017021983.1 | XP_016877472.1 | 2, 3 | 294 | |||
XM_017021984.1 | XP_016877473.1 | 12, 13 | 1563 | |||
GenBank mRNA | AJ315735.1 | - | 13, 14 | 1905 | ||
AK131344.1 | - | 12, 13 | 1534 |