Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015338.5 | NP_056153.2 | 13 | 4172 | ||
XM_006723727.3 | XP_006723790.1 | 12 | 4451 | |||
XM_006723728.3 | XP_006723791.1 | 13 | 3781 | |||
XM_006723730.3 | XP_006723793.1 | 13 | 3977 | |||
XM_006723732.2 | XP_006723795.1 | 12 | 3800 | |||
XM_006723733.1 | XP_006723796.1 | 6 | 3806 | |||
XM_011528648.2 | XP_011526950.1 | 13 | 4096 | |||
XM_011528652.2 | XP_011526954.1 | 14 | 5114 | |||
XM_017027704.1 | XP_016883193.1 | 13 | 5237 | |||
XM_017027705.1 | XP_016883194.1 | 13 | 4250 | |||
XM_017027706.1 | XP_016883195.1 | 12 | 4301 | |||
GenBank mRNA | AB023195.2 | - | 7 | 3222 | ||
AB084280.1 | - | 12 | 3725 | |||
AJ438952.2 | - | 13 | 3998 | |||
AK025756.1 | - | 1 | 118 | |||
AK122923.1 | - | 13 | 3746 | |||
AL117518.1 | - | 6 | 3142 | |||
AL117647.1 | - | 1 | 1105 | |||
BC137278.1 | - | 14 | 3935 | 9021275 | ||
BC137280.1 | - | 14 | 3935 | 9021277 |