Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128933.2 | NP_001122405.1 | 4 | 2427 | ||
NM_001128934.2 | NP_001122406.1 | 4 | 2427 | |||
NM_001286754.1 | NP_001273683.1 | 4 | 2379 | |||
NM_133477.2 | NP_597734.2 | 4 | 2427 | |||
GenBank mRNA | AJ010482.1 | - | 2 | 1447 | ||
AK296962.1 | - | 2 | 1193 | |||
AK307711.1 | - | 4 | 2423 | |||
AL832031.1 | - | 4 | 2410 | |||
AL832363.1 | - | 4 | 2321 | |||
AL833294.1 | - | 4 | 2413 | |||
AL833547.1 | - | 4 | 2427 | |||
BC136256.1 | - | 4 | 2302 | 9020243 | ||
BC150629.1 | - | 4 | 2426 | 9056999 | ||
BX647849.1 | - | 2 | 1936 | |||
BX648090.1 | - | 3 | 2085 | |||
CR936673.1 | - | 4 | 2419 | |||
EU481975.1 | - | 4 | 2231 | |||
FN422000.1 | - | 2 | 1482 | |||
KF147165.1 | - | 4 | 2379 |