Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018050.3 | NP_001018060.1 | 1 | 464 | ||
NM_001018052.3 | NP_001018062.1 | 1 | 464 | |||
NM_001282884.1 | NP_001269813.1 | 2 | 192 | |||
NM_001282885.1 | NP_001269814.1 | 2 | 151 | |||
NM_138338.4 | NP_612211.1 | 2 | 158 | |||
XM_011529993.1 | XP_011528295.1 | 1 | 372 | |||
XM_017028641.1 | XP_016884130.1 | 1 | 372 | |||
GenBank mRNA | AK026524.1 | - | 2 | 138 | ||
AK129663.1 | - | 2 | 141 | |||
AY092087.1 | - | 2 | 136 | |||
BC017248.2 | - | 1 | 351 | 4892259 | ||
BC088367.1 | - | 1 | 327 | 30385483 | ||
BI754673.1 | - | 1 | 347 | |||
BU173504.1 | - | 1 | 106 | |||
BU928882.1 | - | 469 | ||||
BX331353.2 | - | 1 | 430 | |||
BX367405.2 | - | 1 | 385 | |||
CB126821.1 | - | 1 | 437 | |||
CR456459.1 | - | 2 | 192 | |||
DA238361.1 | - | 1 | 477 | |||
DA449644.1 | - | 2 | 173 | |||
DA504810.1 | - | 2 | 140 |