Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308256.1 | NP_001295185.1 | 2 | 369 | ||
NM_138340.4 | NP_612213.2 | 2 | 369 | |||
XM_011525847.2 | XP_011524149.1 | 2 | 342 | |||
XM_017025571.1 | XP_016881060.1 | 2 | 345 | |||
XM_017025572.1 | XP_016881061.1 | 2 | 219 | |||
XM_017025573.1 | XP_016881062.1 | 2 | 129 | |||
GenBank mRNA | AF007152.1 | - | 2 | 328 | ||
AK024880.1 | - | 2 | 243 | |||
BC021196.2 | - | 2 | 329 | 3942164 | ||
BI918326.1 | - | 2 | 321 | |||
DB126083.1 | - | 2 | 369 |