Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308256.1 | NP_001295185.1 | 7 | 1090 | ||
NM_001308257.1 | NP_001295186.1 | 5 | 978 | |||
NM_138340.4 | NP_612213.2 | 9 | 1249 | |||
XM_017025571.1 | XP_016881060.1 | 9 | 1665 | |||
XM_017025572.1 | XP_016881061.1 | 8 | 1493 | |||
XM_017025573.1 | XP_016881062.1 | 10 | 1474 | |||
XM_017025574.1 | XP_016881063.1 | 8 | 1417 | |||
GenBank mRNA | AF007152.1 | - | 9 | 1208 | ||
AK024880.1 | - | 9 | 1123 | |||
AK024992.1 | - | 5 | 458 | |||
BC021196.2 | - | 9 | 1209 | 3942164 | ||
BG547174.1 | - | 5 | 590 |