Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019016.2 | NP_061889.2 | 4 | 990 | ||
XM_006721739.3 | XP_006721802.1 | 5 | 681 | |||
XM_011524460.2 | XP_011522762.1 | 5 | 666 | |||
XM_017024299.1 | XP_016879788.1 | 4 | 975 | |||
GenBank mRNA | AK000268.1 | - | 4 | 990 | ||
BC111968.1 | - | 4 | 965 | 8327436 |