Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178111.1 | NP_001171582.1 | 8 | 1448 | ||
NM_001178112.1 | NP_001171583.1 | 8 | 1415 | |||
NM_138967.3 | NP_620417.1 | 7 | 1365 | |||
NR_033660.1 | 6 | 1208 | ||||
XM_006720420.1 | XP_006720483.1 | 7 | 1524 | |||
XM_006720421.1 | XP_006720484.1 | 8 | 1406 | |||
GenBank mRNA | AB105192.1 | - | 7 | 1336 | ||
AF495715.1 | - | 7 | 1317 | |||
AK025678.1 | - | 1 | 174 | |||
AK056438.1 | - | 8 | 1380 | |||
AK126940.1 | - | 7 | 1365 | |||
AK127480.1 | - | 8 | 1385 | |||
AK301505.1 | - | 6 | 1170 | |||
AL833230.1 | - | 1 | 531 | |||
AL834226.1 | - | 7 | 1362 | |||
BC024700.1 | - | 6 | 1208 | 4941050 | ||
CR936761.1 | - | 7 | 1298 |