Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153698.1 | NP_714542.1 | 6 | 634 | ||
XM_005251783.3 | XP_005251840.1 | 5 | 497 | |||
XM_005251784.4 | XP_005251841.1 | 6 | 486 | |||
GenBank mRNA | BC136501.1 | - | 6 | 472 | 9020490 | |
BC136503.1 | - | 6 | 634 | 9020492 | ||
BF445089.1 | - | 6 | 509 | |||
BM843088.1 | - | 5 | 502 |