Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030059.2 | NP_001025230.1 | 2 | 489 | ||
NM_001318166.1 | NP_001305095.1 | 2 | 489 | |||
NM_001318167.1 | NP_001305096.1 | 2 | 489 | |||
NM_001318168.1 | NP_001305097.1 | 2 | 489 | |||
NM_001318169.1 | NP_001305098.1 | 2 | 489 | |||
NR_134516.1 | 3 | 281 | ||||
XM_005269592.2 | XP_005269649.1 | 2 | 489 | |||
XM_006717685.3 | XP_006717748.1 | 2 | 158 | |||
XM_006717686.3 | XP_006717749.1 | 2 | 489 | |||
XM_011539444.2 | XP_011537746.1 | 3 | 216 | |||
XM_011539445.2 | XP_011537747.1 | 2 | 489 | |||
XM_017015820.1 | XP_016871309.1 | 2 | 161 | |||
XM_017015823.1 | XP_016871312.1 | 2 | 489 | |||
XM_017015824.1 | XP_016871313.1 | 2 | 489 | |||
GenBank mRNA | AK098668.1 | - | 2 | 107 | ||
AK296273.1 | - | 2 | 196 | |||
AK310178.1 | - | 2 | 140 | |||
BC101267.2 | - | 3 | 287 | |||
BC101268.2 | - | 3 | 183 | 40024099 | ||
BC101269.1 | - | 3 | 164 | |||
BC122535.1 | - | 3 | 286 | |||
BC132787.1 | - | 2 | 312 | 40146809 | ||
BI550027.1 | - | 2 | 221 | |||
DB473853.2 | - | 2 | 196 | |||
DB495310.2 | - | 2 | 466 | |||
DR000933.1 | - | 2 | 420 |