Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113528.1 | NP_001107000.1 | 4, 5 | 860 | ||
NM_001297775.1 | NP_001284704.1 | 4, 5 | 860 | |||
NM_152636.2 | NP_689849.2 | 4, 5 | 860 | |||
XM_011519935.2 | XP_011518237.1 | 6, 7 | 1081 | |||
XM_011519937.2 | XP_011518239.1 | 3, 4 | 322 | |||
XM_011519940.2 | XP_011518242.1 | 4, 5 | 860 | |||
XM_017017297.1 | XP_016872786.1 | 4, 5 | 764 | |||
XM_017017298.1 | XP_016872787.1 | 7, 8 | 1120 | |||
XM_017017299.1 | XP_016872788.1 | 3, 4 | 1609 | |||
XM_017017300.1 | XP_016872789.1 | 5, 6 | 817 | |||
XM_017017301.1 | XP_016872790.1 | 4, 5 | 860 | |||
GenBank mRNA | AK091298.1 | - | 3, 4 | 512 | ||
AL832668.1 | - | 3, 4 | 513 | |||
BC030997.1 | - | 3, 4 | 533 | 4657323 | ||
BC066900.1 | - | 4, 5 | 742 |