Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145058.2 | NP_659495.1 | 4 | 1293 | ||
NR_130703.1 | 3 | 1141 | ||||
GenBank mRNA | AB085763.1 | - | 4 | 1056 | ||
AK054942.1 | - | 1 | 81 | |||
AK056934.1 | - | 4 | 1277 | |||
BC013042.2 | - | 4 | 1066 | 3544761 |