Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41 SIRNA Assays ›
Gene Symbol:
EPB41
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
4.1R, EL1, HE
Chromosome Location:
Chr.1: 28887091 - 29120046 on Build GRCh38
UniGene ID:
Hs.175437
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4701
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166005.1
NP_001159477.1
10
1538
NM_001166006.1
NP_001159478.1
10
1611
NM_001166007.1
NP_001159479.1
10
1521
NM_004437.3
NP_004428.1
11
1601
NM_203342.2
NP_976217.1
11
1618
NM_203343.2
NP_976218.1
9
1433
XM_005245753.1
XP_005245810.1
10
1613
XM_005245757.1
XP_005245814.1
10
1613
XM_005245760.1
XP_005245817.1
10
1613
XM_005245761.1
XP_005245818.1
10
1613
XM_005245763.1
XP_005245820.1
10
1613
XM_005245764.1
XP_005245821.1
10
1613
XM_005245765.1
XP_005245822.1
10
1613
XM_005245768.1
XP_005245825.1
10
1613
XM_005245769.1
XP_005245826.1
10
1613
XM_005245770.1
XP_005245827.1
10
1613
XM_005245772.4
XP_005245829.1
10
1613
XM_005245773.4
XP_005245830.1
10
1613
XM_005245774.1
XP_005245831.1
10
1613
XM_006710434.1
XP_006710497.1
10
1613
XM_006710439.1
XP_006710502.1
10
1613
XM_011540956.1
XP_011539258.1
10
1613
XM_011540957.1
XP_011539259.1
10
1613
XM_011540958.1
XP_011539260.1
10
1613
XM_011540959.1
XP_011539261.1
10
1613
XM_011540960.1
XP_011539262.1
10
1613
XM_011540961.1
XP_011539263.1
10
1613
XM_011540962.1
XP_011539264.1
10
1613
XM_011540963.2
XP_011539265.1
10
1613
XM_011540964.1
XP_011539266.1
10
1509
XM_011540965.2
XP_011539267.1
10
1613
XM_017000581.1
XP_016856070.1
9
1508
XM_017000582.1
XP_016856071.1
9
1508
XM_017000583.1
XP_016856072.1
10
1613
XM_017000584.1
XP_016856073.1
10
1904
XM_017000585.1
XP_016856074.1
10
1901
XM_017000586.1
XP_016856075.1
10
1613
XM_017000587.1
XP_016856076.1
10
1613
XM_017000588.1
XP_016856077.1
10
1613
XM_017000589.1
XP_016856078.1
9
1508
XM_017000590.1
XP_016856079.1
9
1508
XM_017000591.1
XP_016856080.1
10
1613
XM_017000592.1
XP_016856081.1
10
1613
XM_017000593.1
XP_016856082.1
10
1509
XM_017000594.1
XP_016856083.1
10
1509
XM_017000595.1
XP_016856084.1
10
1509
XM_017000596.1
XP_016856085.1
10
1509
XM_017000597.1
XP_016856086.1
10
1509
XM_017000598.1
XP_016856087.1
10
1509
XM_017000599.1
XP_016856088.1
10
1509
XM_017000600.1
XP_016856089.1
10
1509
XM_017000601.1
XP_016856090.1
10
1509
XM_017000602.1
XP_016856091.1
10
1509
XM_017000603.1
XP_016856092.1
10
1509
XM_017000604.1
XP_016856093.1
10
1509
GenBank mRNA
AB209649.1
-
10
1521
AF156225.1
-
9
1411
AK310655.1
-
9
1449
BC039079.1
-
10
1608
5167140
BC096104.1
-
7
827
BC096105.1
-
9
1145
40001729
BC113841.1
-
9
1145
BC114450.1
-
9
1145
J03796.1
-
9
1352
M14993.1
-
11
1582
M61733.1
-
11
1599
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
blood circulation
actin cytoskeleton organization
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural constituent of cytoskeleton
protein binding
calmodulin binding
1-phosphatidylinositol binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0