Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYRFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182530.2 | NP_872336.2 | 21 | 2500 | ||
XM_011539052.2 | XP_011537354.1 | 20 | 2981 | |||
XM_011539055.1 | XP_011537357.1 | 19 | 1968 | |||
XM_017018967.1 | XP_016874456.1 | 20 | 2782 | |||
XM_017018968.1 | XP_016874457.1 | 21 | 3236 | |||
XM_017018969.1 | XP_016874458.1 | 20 | 2716 | |||
XM_017018970.1 | XP_016874459.1 | 19 | 2526 | |||
XM_017018973.1 | XP_016874462.1 | 17 | 2197 | |||
GenBank mRNA | AK057785.1 | - | 8 | 617 | ||
BC101820.1 | - | 6 | 452 | |||
BC143553.1 | - | 6 | 452 | 9052064 |