Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB41L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258329.1 | NP_001245258.1 | 16 | 2005 | ||
NM_001258331.1 | NP_001245260.1 | 14 | 1731 | |||
NM_012156.2 | NP_036288.2 | 15 | 1951 | |||
NM_177996.2 | NP_818932.1 | 14 | 1726 | |||
XM_011528667.1 | XP_011526969.1 | 22 | 4934 | |||
XM_011528671.1 | XP_011526973.1 | 18 | 4021 | |||
XM_011528673.1 | XP_011526975.1 | 17 | 3981 | |||
XM_011528677.1 | XP_011526979.1 | 17 | 3795 | |||
XM_011528681.1 | XP_011526983.1 | 13 | 1758 | |||
XM_011528682.1 | XP_011526984.1 | 13 | 1755 | |||
XM_011528683.1 | XP_011526985.1 | 14 | 1794 | |||
XM_017027711.1 | XP_016883200.1 | 16 | 3744 | |||
XM_017027712.1 | XP_016883201.1 | 15 | 1845 | |||
XM_017027713.1 | XP_016883202.1 | 14 | 1788 | |||
XM_017027714.1 | XP_016883203.1 | 14 | 1785 | |||
XM_017027715.1 | XP_016883204.1 | 13 | 1755 | |||
XM_017027716.1 | XP_016883205.1 | 16 | 1984 | |||
XM_017027717.1 | XP_016883206.1 | 15 | 1942 | |||
GenBank mRNA | AB002336.1 | - | 15 | 1941 | ||
AK096848.1 | - | 14 | 1778 | |||
AK299817.1 | - | 15 | 1898 | |||
AY049789.1 | - | 14 | 1789 | |||
BC013885.2 | - | 14 | 1731 | 3688606 | ||
BC040259.1 | - | 16 | 1983 | 5195986 | ||
BX537978.1 | - | 5 | 2370 |