Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198568.1 | NP_001185497.1 | 5 | 1204 | ||
NM_001198592.1 | NP_001185521.1 | 6 | 903 | |||
NM_139247.3 | NP_640340.2 | 6 | 899 | |||
GenBank mRNA | AF497516.1 | - | 5 | 791 | ||
AK122714.1 | - | 6 | 899 | |||
AK126468.1 | - | 5 | 753 | |||
BC117473.1 | - | 5 | 880 | 40126024 | ||
BC117475.1 | - | 5 | 880 | 40126026 | ||
BC143947.1 | - | 5 | 880 | |||
BX248285.1 | - | 6 | 870 | |||
BX537489.1 | - | 1 | 1244 |