Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012642.2 | NP_001012660.1 | 10 | 886 | ||
XM_011521327.2 | XP_011519629.1 | 10 | 995 | |||
XM_011521328.2 | XP_011519630.1 | 10 | 1346 | |||
XM_011521329.2 | XP_011519631.1 | 10 | 863 | |||
XM_011521330.2 | XP_011519632.1 | 10 | 947 | |||
XM_011521331.2 | XP_011519633.1 | 9 | 829 | |||
XM_011521332.2 | XP_011519634.1 | 11 | 1070 | |||
XM_011521333.2 | XP_011519635.1 | 11 | 1087 | |||
XM_017021997.1 | XP_016877486.1 | 10 | 1030 | |||
XM_017021998.1 | XP_016877487.1 | 10 | 1254 | |||
GenBank mRNA | AK095072.1 | - | 10 | 876 | ||
AK096501.1 | - | 1 | 405 | |||
AK303724.1 | - | 2 | 294 | |||
BC038451.1 | - | 10 | 860 |