Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 9 | 809 | ||
XM_006721162.1 | XP_006721225.1 | 8 | 739 | |||
XM_011522928.1 | XP_011521230.1 | 9 | 987 | |||
XM_011522929.1 | XP_011521231.1 | 9 | 852 | |||
XM_011522930.2 | XP_011521232.1 | 9 | 820 | |||
XM_011522931.2 | XP_011521233.1 | 9 | 820 | |||
XM_017023012.1 | XP_016878501.1 | 9 | 836 | |||
XM_017023013.1 | XP_016878502.1 | 7 | 804 | |||
XM_017023014.1 | XP_016878503.1 | 5 | 936 | |||
GenBank mRNA | AJ441184.1 | - | 9 | 808 | ||
AK056456.1 | - | 9 | 808 | |||
AK074168.1 | - | 8 | 1226 | |||
AK092025.1 | - | 2 | 125 | |||
AK096727.1 | - | 1 | 15 | |||
AK128387.1 | - | 9 | 905 | |||
AK315590.1 | - | 9 | 812 | |||
BC013735.1 | - | 7 | 595 | |||
BC032542.1 | - | 9 | 815 | 5534376 |