Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 7 | 606 | ||
XM_006721162.1 | XP_006721225.1 | 7 | 617 | |||
XM_011522928.1 | XP_011521230.1 | 7 | 784 | |||
XM_011522929.1 | XP_011521231.1 | 7 | 649 | |||
XM_011522930.2 | XP_011521232.1 | 7 | 617 | |||
XM_011522931.2 | XP_011521233.1 | 7 | 617 | |||
XM_017023012.1 | XP_016878501.1 | 7 | 633 | |||
XM_017023014.1 | XP_016878503.1 | 3 | 733 | |||
GenBank mRNA | AJ441184.1 | - | 7 | 605 | ||
AK056456.1 | - | 7 | 605 | |||
AK074168.1 | - | 6 | 1023 | |||
AK128387.1 | - | 7 | 702 | |||
AK315590.1 | - | 7 | 609 | |||
BC013735.1 | - | 5 | 392 | |||
BC032542.1 | - | 7 | 612 | 5534376 |