Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000476.2 | NP_000467.1 | 7 | 2077 | ||
NM_001318121.1 | NP_001305050.1 | 7 | 2064 | |||
NM_001318122.1 | NP_001305051.1 | 6 | 2072 | |||
GenBank mRNA | EU794628.1 | - | 7 | 2077 |