Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000119.2 | NP_000110.2 | 10 | 1940 | ||
NM_001114134.1 | NP_001107606.1 | 10 | 1850 | |||
XM_005254225.1 | XP_005254282.1 | 9 | 1582 | |||
XM_011521349.2 | XP_011519651.1 | 11 | 2668 | |||
XM_011521350.2 | XP_011519652.1 | 12 | 2400 | |||
XM_011521351.2 | XP_011519653.1 | 12 | 2262 | |||
XM_011521352.2 | XP_011519654.1 | 10 | 1843 | |||
XM_011521353.2 | XP_011519655.1 | 9 | 1773 | |||
XM_011521354.2 | XP_011519656.1 | 9 | 1254 | |||
GenBank mRNA | AK297146.1 | - | 10 | 1624 | ||
AK297663.1 | - | 9 | 1500 | |||
BC096093.4 | - | 10 | 1629 | 40001236 | ||
BC096094.2 | - | 10 | 1629 | 40001240 | ||
BC096095.4 | - | 5 | 1064 | |||
BC099627.1 | - | 9 | 1413 | 40001239 | ||
M29399.1 | - | 10 | 1737 | |||
M30646.1 | - | 10 | 1777 | |||
M30647.1 | - | 10 | 1867 | |||
M60298.1 | - | 10 | 2099 |