Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001214906.1 | NP_001201835.1 | 3 | 231 | ||
NM_001214907.1 | NP_001201836.1 | 2 | 135 | |||
NM_001214909.1 | NP_001201838.1 | 3 | 599 | |||
NM_001324494.1 | NP_001311423.1 | 2 | 505 | |||
NM_152652.2 | NP_689865.2 | 2 | 493 | |||
GenBank mRNA | AK056313.1 | - | 2 | 176 | ||
BC041388.1 | - | 2 | 180 | 5276303 | ||
BU854776.1 | - | 3 | 568 | |||
CF594347.1 | - | 296 | ||||
DA492655.1 | - | 2 | 135 | |||
DA508718.1 | - | 3 | 233 |