Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174918.2 | NP_777578.2 | 5, 6 | 509 | ||
GenBank mRNA | AF461155.1 | - | 5, 6 | 938 | ||
AF461156.1 | - | 5, 6 | 488 | |||
BC035847.1 | - | 5, 6 | 451 | |||
BC104796.1 | - | 5, 6 | 509 | 8143799 | ||
BC104798.1 | - | 5, 6 | 509 | 8143801 |