Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000119.2 | NP_000110.2 | 13 | 2420 | ||
NM_001114134.1 | NP_001107606.1 | 13 | 2330 | |||
XM_005254225.1 | XP_005254282.1 | 12 | 2062 | |||
XM_011521349.2 | XP_011519651.1 | 14 | 3148 | |||
XM_011521350.2 | XP_011519652.1 | 15 | 2880 | |||
XM_011521351.2 | XP_011519653.1 | 15 | 2742 | |||
XM_011521352.2 | XP_011519654.1 | 13 | 2323 | |||
XM_011521353.2 | XP_011519655.1 | 12 | 2253 | |||
XM_011521354.2 | XP_011519656.1 | 12 | 1734 | |||
GenBank mRNA | AK297146.1 | - | 13 | 2104 | ||
AK297663.1 | - | 12 | 1980 | |||
BC096093.4 | - | 13 | 2109 | 40001236 | ||
BC096094.2 | - | 13 | 2109 | 40001240 | ||
BC096095.4 | - | 8 | 1544 | |||
BC099627.1 | - | 12 | 1893 | 40001239 | ||
M29399.1 | - | 13 | 2217 | |||
M30646.1 | - | 13 | 2257 | |||
M30647.1 | - | 13 | 2347 | |||
M60298.1 | - | 13 | 2579 |