Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CADM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145296.1 | NP_660339.1 | 5 | 611 | ||
XM_005258620.2 | XP_005258677.1 | 5 | 593 | |||
XM_017026452.1 | XP_016881941.1 | 5 | 520 | |||
GenBank mRNA | AF363368.1 | - | 5 | 611 | ||
AK313028.1 | - | 5 | 593 | |||
BC068457.1 | - | 5 | 592 | |||
BC113963.1 | - | 2 | 184 |