Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010866.3 | NP_001010866.1 | 4 | 573 | ||
NM_001130924.2 | NP_001124396.2 | 4 | 573 | |||
XM_006710417.3 | XP_006710480.1 | 4 | 604 | |||
XM_011540910.2 | XP_011539212.1 | 4 | 602 | |||
XM_017000549.1 | XP_016856038.1 | 4 | 604 | |||
XM_017000550.1 | XP_016856039.1 | 4 | 604 | |||
XM_017000551.1 | XP_016856040.1 | 4 | 814 | |||
XM_017000552.1 | XP_016856041.1 | 4 | 642 | |||
GenBank mRNA | BC121187.1 | - | 4 | 528 | 3870153 | |
BC137291.1 | - | 4 | 573 | 9021288 | ||
BX648377.1 | - | 2 | 244 |