Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256946.1 | NP_001243875.1 | 2 | 193 | ||
NM_001282670.1 | NP_001269599.1 | 6 | 3039 | |||
NM_001282671.1 | NP_001269600.1 | 2 | 193 | |||
NM_001282672.1 | NP_001269601.1 | 2 | 193 | |||
NM_001282673.1 | NP_001269602.1 | 2 | 193 | |||
NM_182533.2 | NP_872339.2 | 2 | 193 | |||
XM_006710419.3 | XP_006710482.1 | 3 | 778 | |||
XM_011540914.2 | XP_011539216.1 | 7 | 729 | |||
XM_011540921.2 | XP_011539223.1 | 4 | 578 | |||
XM_011540922.1 | XP_011539224.1 | 2 | 192 | |||
XM_017000553.1 | XP_016856042.1 | 5 | 576 | |||
GenBank mRNA | AF461903.1 | - | 2 | 2007 | ||
AF469129.1 | - | 2 | 159 | |||
AK055593.1 | - | 2 | 193 | |||
AK126870.1 | - | 6 | 3038 | |||
BC041041.1 | - | 2 | 132 | |||
BC066360.1 | - | 2 | 190 | |||
BC103992.2 | - | 2 | 193 | 40022365 | ||
BG825658.1 | - | 2 | 173 | |||
BU542153.1 | - | 1 | 18 | |||
DA596011.1 | - | 2 | 193 |