Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011547.2 | NP_001011547.2 | 12 | 1712 | ||
NM_001135181.1 | NP_001128653.1 | 13 | 1787 | |||
XM_011540924.2 | XP_011539226.1 | 12 | 3393 | |||
XM_011540925.2 | XP_011539227.1 | 12 | 3472 | |||
XM_011540926.2 | XP_011539228.1 | 11 | 1973 | |||
XM_011540927.1 | XP_011539229.1 | 10 | 1571 | |||
XM_011540928.1 | XP_011539230.1 | 12 | 1591 | |||
GenBank mRNA | AK131200.1 | - | 12 | 1665 | ||
AK311580.1 | - | 1 | 142 | |||
BC104857.1 | - | 12 | 1712 | 8143860 | ||
BC104863.1 | - | 12 | 1712 | 8143866 | ||
BC143570.1 | - | 13 | 1775 | 9052081 | ||
BC143572.1 | - | 13 | 1785 | 9052083 | ||
BC143587.1 | - | 11 | 1506 | |||
BX648549.1 | - | 13 | 1741 |