Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011547.2 | NP_001011547.2 | 7 | 849 | ||
NM_001135181.1 | NP_001128653.1 | 8 | 924 | |||
XM_011540924.2 | XP_011539226.1 | 7 | 2608 | |||
XM_011540925.2 | XP_011539227.1 | 7 | 2609 | |||
XM_011540926.2 | XP_011539228.1 | 6 | 1110 | |||
XM_011540927.1 | XP_011539229.1 | 5 | 708 | |||
XM_011540928.1 | XP_011539230.1 | 7 | 728 | |||
XM_011540929.2 | XP_011539231.1 | 7 | 2613 | |||
XM_017000558.1 | XP_016856047.1 | 7 | 2612 |