Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 22 | 2969 | ||
NM_183372.5 | NP_899228.4 | 24 | 3663 | |||
NR_046188.3 | 25 | 3731 | ||||
NR_110864.1 | 23 | 3655 | ||||
GenBank mRNA | AK054850.1 | - | 8 | 1412 | ||
AK302413.1 | - | 21 | 2805 | |||
AL832622.2 | - | 25 | 3866 | |||
BC136292.1 | - | 24 | 3663 | 9020279 | ||
BX648497.1 | - | 23 | 3454 |
Gene Symbol: NBPF13P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103466.1 | 2 | 189 | |||
GenBank mRNA | BX336604.2 | - | 2 | 189 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY894561.1 | - | 13 | 1623 | ||
AY894571.1 | - | 19 | 2713 |