Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 22 | 3390 | ||
NM_183372.5 | NP_899228.4 | 24 | 4084 | |||
NR_046188.3 | 25 | 4152 | ||||
NR_110864.1 | 23 | 4076 | ||||
GenBank mRNA | AK054850.1 | - | 8 | 1833 | ||
AL832622.2 | - | 25 | 4287 | |||
BX648497.1 | - | 23 | 3875 |
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094705.1 | - | 14 | 2096 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095459.1 | - | 12 | 2536 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 17 | 2529 | ||
GenBank mRNA | AK292666.1 | - | 8 | 1282 | ||
CR936830.1 | - | 5 | 3942 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 20 | 3241 | ||
NR_102404.1 | 19 | 3077 | ||||
NR_102405.1 | 18 | 2974 | ||||
GenBank mRNA | BC086308.1 | - | 20 | 3084 | 5453529 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027348.1 | - | 1 | 149 |