Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTEX1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152665.2 | NP_689878.2 | 3 | 315 | ||
XM_011540933.2 | XP_011539235.1 | 4 | 702 | |||
XM_017000566.1 | XP_016856055.1 | 3 | 611 | |||
GenBank mRNA | AK098192.1 | - | 3 | 313 | ||
AK307100.1 | - | 3 | 277 | |||
AK307878.1 | - | 3 | 274 | |||
BC106885.2 | - | 3 | 280 | 40029630 | ||
DA520308.1 | - | 3 | 315 |