Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTCAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173852.3 | NP_776251.1 | 5 | 408 | ||
GenBank mRNA | AK024167.1 | - | 4 | 3125 | ||
AK311907.1 | - | 5 | 382 | |||
AY157577.1 | - | 4 | 293 | |||
BC029806.1 | - | 5 | 395 | 5189880 | ||
BC048205.1 | - | 5 | 334 | |||
BC057233.1 | - | 5 | 417 | 6463372 | ||
BX647396.1 | - | 4 | 2732 |