Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005232.4 | NP_005223.4 | 14 | 2312 | ||
XM_006715880.3 | XP_006715943.1 | 13 | 1940 | |||
GenBank mRNA | AK290351.1 | - | 14 | 2276 | ||
AK313263.1 | - | 14 | 2285 | |||
BC130291.1 | - | 14 | 2296 | 40146322 | ||
M18391.1 | - | 14 | 2342 | |||
Z27409.1 | - | 10 | 1370 |