Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181715.2 | NP_859066.1 | 11 | 1514 | ||
XM_005244946.1 | XP_005245003.1 | 11 | 1510 | |||
XM_005244949.2 | XP_005245006.1 | 11 | 1387 | |||
XM_017000575.1 | XP_016856064.1 | 10 | 1283 | |||
XM_017000576.1 | XP_016856065.1 | 10 | 1268 | |||
XM_017000577.1 | XP_016856066.1 | 10 | 1412 | |||
XM_017000578.1 | XP_016856067.1 | 10 | 1397 | |||
XM_017000579.1 | XP_016856068.1 | 5 | 809 | |||
GenBank mRNA | AF289574.1 | - | 2 | 473 | ||
AK223559.1 | - | 11 | 1467 | |||
AY360172.1 | - | 11 | 1485 | |||
AY726597.1 | - | 11 | 1294 | |||
BC033304.1 | - | 9 | 1116 | |||
BC053562.1 | - | 11 | 1425 | 6188068 |