Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144992.4 | NP_659429.4 | 26 | 3744 | ||
XM_005263897.2 | XP_005263954.1 | 26 | 3709 | |||
XM_006712357.2 | XP_006712420.1 | 25 | 3458 | |||
XM_006712359.3 | XP_006712422.1 | 17 | 2138 | |||
XM_006712360.3 | XP_006712423.1 | 14 | 1785 | |||
XM_011510770.1 | XP_011509072.1 | 25 | 3658 | |||
XM_011510771.2 | XP_011509073.1 | 25 | 3583 | |||
XM_011510772.1 | XP_011509074.1 | 25 | 3571 | |||
XM_017003560.1 | XP_016859049.1 | 20 | 2575 | |||
XM_017003561.1 | XP_016859050.1 | 20 | 2581 | |||
XM_017003562.1 | XP_016859051.1 | 20 | 2631 | |||
XM_017003563.1 | XP_016859052.1 | 20 | 2581 | |||
GenBank mRNA | AK090786.1 | - | 13 | 1580 | ||
AK093084.1 | - | 14 | 1711 | |||
AK098841.1 | - | 8 | 983 | |||
AK295743.1 | - | 20 | 2639 | |||
AL832761.1 | - | 27 | 3915 | |||
AL834173.1 | - | 17 | 2143 | |||
AL834385.1 | - | 16 | 2007 | |||
BC022028.2 | - | 8 | 974 | |||
BC150653.1 | - | 26 | 3684 | 9057024 |