Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287491.1 | NP_001274420.1 | 3, 4 | 2492 | ||
XM_005264187.3 | XP_005264244.1 | 4, 5 | 3012 | |||
XM_011532682.2 | XP_011530984.1 | 4, 5 | 3012 | |||
XM_011532683.2 | XP_011530985.1 | 4, 5 | 3321 | |||
XM_011532684.2 | XP_011530986.1 | 4, 5 | 4753 | |||
XM_011532685.2 | XP_011530987.1 | 3, 4 | 2955 | |||
XM_011532686.2 | XP_011530988.1 | 3, 4 | 2955 | |||
XM_011532687.2 | XP_011530989.1 | 3, 4 | 2294 | |||
XM_011532688.2 | XP_011530990.1 | 4, 5 | 3012 | |||
XM_017003566.1 | XP_016859055.1 | 4, 5 | 4852 | |||
XM_017003567.1 | XP_016859056.1 | 3, 4 | 2324 | |||
GenBank mRNA | AF466365.1 | - | 1, 2 | 1250 | ||
HQ220209.1 | - | 3, 4 | 2492 |