Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173545.2 | NP_775816.1 | 10 | 1562 | ||
GenBank mRNA | AK290841.1 | - | 10 | 1559 | ||
BC030711.2 | - | 7 | 873 | |||
BC041144.1 | - | 10 | 1511 | 6042653 |
Gene Symbol: LINC01323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG192479.1 | - | 59 |