Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC51A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152672.5 | NP_689885.4 | 9 | 1146 | ||
GenBank mRNA | AK172837.1 | - | 9 | 1149 | ||
AY194243.1 | - | 9 | 967 | |||
BC029606.1 | - | 5 | 758 | 5297443 |