Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037325.2 | NP_001032402.1 | 2 | 444 | ||
NM_001199165.2 | NP_001186094.1 | 22 | 2575 | |||
NM_001302891.1 | NP_001289820.1 | 20 | 2525 | |||
XM_005257119.4 | XP_005257176.1 | 22 | 2669 | |||
XM_005257125.3 | XP_005257182.1 | 14 | 1659 | |||
XM_005257126.4 | XP_005257183.1 | 11 | 1324 | |||
XM_006721740.2 | XP_006721803.1 | 22 | 2541 | |||
XM_011524461.2 | XP_011522763.1 | 22 | 2682 | |||
XM_011524462.2 | XP_011522764.1 | 22 | 2671 | |||
XM_011524463.2 | XP_011522765.1 | 22 | 2671 | |||
XM_011524465.2 | XP_011522767.1 | 19 | 1998 | |||
XM_011524466.2 | XP_011522768.1 | 12 | 1477 | |||
XM_011524467.2 | XP_011522769.1 | 11 | 1319 | |||
XM_017024302.1 | XP_016879791.1 | 20 | 2412 | |||
XM_017024303.1 | XP_016879792.1 | 23 | 2801 | |||
GenBank mRNA | AF458591.1 | - | 12 | 1482 | ||
AK097612.1 | - | 4 | 543 | |||
AK299075.1 | - | 22 | 2526 | |||
BC029524.1 | - | 2 | 444 | |||
BC131805.1 | - | 20 | 2353 | 40119069 | ||
BX097629.1 | - | 2 | 122 | |||
CX872964.1 | - | 6 | 768 |