Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037325.2 | NP_001032402.1 | 3, 4 | 620 | ||
NM_001199165.2 | NP_001186094.1 | 23, 24 | 2751 | |||
NM_001302891.1 | NP_001289820.1 | 21, 22 | 2701 | |||
XM_005257119.4 | XP_005257176.1 | 23, 24 | 2845 | |||
XM_005257125.3 | XP_005257182.1 | 15, 16 | 1835 | |||
XM_005257126.4 | XP_005257183.1 | 12, 13 | 1500 | |||
XM_006721740.2 | XP_006721803.1 | 23, 24 | 2717 | |||
XM_011524461.2 | XP_011522763.1 | 23, 24 | 2858 | |||
XM_011524462.2 | XP_011522764.1 | 23, 24 | 2847 | |||
XM_011524463.2 | XP_011522765.1 | 23, 24 | 2847 | |||
XM_011524465.2 | XP_011522767.1 | 20, 21 | 2174 | |||
XM_011524466.2 | XP_011522768.1 | 13, 14 | 1653 | |||
XM_011524467.2 | XP_011522769.1 | 12, 13 | 1495 | |||
XM_017024302.1 | XP_016879791.1 | 21, 22 | 2588 | |||
GenBank mRNA | AF458591.1 | - | 13, 14 | 1658 | ||
AK097612.1 | - | 5, 6 | 719 | |||
AK299075.1 | - | 23, 24 | 2702 | |||
BC029524.1 | - | 3, 4 | 620 | |||
BC131805.1 | - | 21, 22 | 2529 | 40119069 | ||
BX097629.1 | - | 3, 4 | 298 |