Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105571.2 | NP_001099041.1 | 4 | 575 | ||
NM_001220493.1 | NP_001207422.1 | 4 | 578 | |||
GenBank mRNA | BC147024.1 | - | 4 | 575 | 9056952 | |
BC147025.1 | - | 4 | 575 | 9056953 | ||
BC171909.1 | - | 4 | 578 | 9054563 | ||
DN831947.1 | - | 4 | 502 |