Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181716.2 | NP_859067.2 | 5 | 798 | ||
GenBank mRNA | AF514992.1 | - | 5 | 734 | ||
AY382899.1 | - | 3 | 772 | |||
BC052604.1 | - | 5 | 808 | 5934088 | ||
BC137486.1 | - | 5 | 830 | 9021486 | ||
BC137487.1 | - | 5 | 830 | 9021487 |