Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181716.2 | NP_859067.2 | 4 | 697 | ||
GenBank mRNA | AF514992.1 | - | 4 | 633 | ||
AY382899.1 | - | 2 | 671 | |||
BC052604.1 | - | 4 | 707 | 5934088 | ||
BC137486.1 | - | 4 | 729 | 9021486 | ||
BC137487.1 | - | 4 | 729 | 9021487 |