Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF385C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242704.1 | NP_001229633.1 | 8 | 2201 | ||
XM_006721751.3 | XP_006721814.1 | 8 | 2309 | |||
XM_011524479.2 | XP_011522781.1 | 9 | 3005 | |||
XM_011524482.2 | XP_011522784.1 | 9 | 3002 | |||
XM_011524483.2 | XP_011522785.1 | 9 | 2991 | |||
XM_017024314.1 | XP_016879803.1 | 10 | 3134 | |||
XM_017024315.1 | XP_016879804.1 | 10 | 2969 | |||
XM_017024316.1 | XP_016879805.1 | 10 | 3131 | |||
XM_017024317.1 | XP_016879806.1 | 10 | 3120 | |||
XM_017024318.1 | XP_016879807.1 | 10 | 3118 | |||
XM_017024319.1 | XP_016879808.1 | 9 | 2988 | |||
XM_017024320.1 | XP_016879809.1 | 9 | 2396 | |||
GenBank mRNA | BC080613.1 | - | 4 | 1594 | 5922386 |